1,3,5-Trithian

Lieferant: Thermo Scientific
Gefahr

140520250 140521000
ACRO140520250EA 67.4 EUR
ACRO140520250 ACRO140521000
1,3,5-Trithian
1,3,5-Trithiane

  • Building Block for chemical synthesis

Formel: C₃H₆S₃
Schmelzpunkt: 214…215 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006653
CAS-Nummer: 291-21-4
UN: 2811
ADR: 6.1,III

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Carbon (C) 25.1 to 27.0
Sulfur (S) 67.1 to 72.0

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR