8-Azaguanine

Lieferant: Thermo Scientific
Achtung

Synonyme: 5-Amino-3,6-dihydro-[1,2,3]triazolo[4,5-d]pyrimidin-7-one, 2-Amino-6-hydroxy-8-azapurine, 5-Amino-1,4-dihydro-7H-[1,2,3]triazolo[4,5-d]pyrimidin-7-one, Azaguanine-8, Pathocidine, 8-Azaguanine, Guanazolo, 5-Amino-1H-triazolo[4,5-d]pyrimidin-7-ol

229920010 229920050
ACRO229920010EA 88.4 EUR
ACRO229920010 ACRO229920050
8-Azaguanine
8-Azaguanine

  • Bioactive

Formel: C₄H₄N₆O
Molecular Weight: 152,12 g/mol
Schmelzpunkt: >300 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00056937
CAS-Nummer: 134-58-7

Bestellen

Spezifikation Testergebnisse

Appearance Yellow to light orange Powder
Infrared spectrum Conforms
Loss on drying ≤5 %
HPLC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR