Piceatannol
Lieferant: Thermo Fisher Scientific
Synonyme:
(E)-4-(3,5-Dihydroxystyryl)benzene-1,2-diol
328450010
328452500
ACRO328450010EA
516
EUR
ACRO328450010
ACRO328452500
Piceatannol
Piceatannol
- Bioactive
Formel:
C₁₄H₁₂O₄ Molecular Weight: 244.2427 g/mol Schmelzpunkt: 235 °C (Zers.) Lagertemperatur: Kühlschrank |
MDL:
MFCD00221715 CAS-Nummer: 10083-24-6 |
Spezifikation Testergebnisse
Appearance | Yellow to brown-pink Crystalline powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...