Tetrahydroxydiborane ≥95%

Lieferant: Thermo Scientific
Achtung

Synonyme: Tetrahydroxydiboron, Hypodiboric Acid, Diboronic Acid

461280050 461280250
ACRO461280050EA 81.3 EUR
ACRO461280050 ACRO461280250
Tetrahydroxydiborane ≥95%
Tetrahydroxydiborane

  • Reagent for chemical synthesis
  • Can be used to prepare boronic acids

Formel: H₄B₂O₄
Molecular Weight: 89,65 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05663888
CAS-Nummer: 13675-18-8

Bestellen

Spezifikation Testergebnisse

Appearance White to light brown Powder or crystals
Infrared spectrum Conforms
GC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR