Betaxolol-hydrochlorid
Lieferant: Thermo Fisher Scientific
|
Achtung
|
462440010
462440050
ACRO462440010EA
108
EUR
ACRO462440010
ACRO462440050
Betaxolol-hydrochlorid
Betaxolol hydrochloride
- Bioactive
Formel:
C₁₈H₂₉NO₃ Schmelzpunkt: 115 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD00242959 CAS-Nummer: 63659-19-8 |
Spezifikation Testergebnisse
Appearance | White to almost Powder |
Infrared spectrum | Conforms |
Melting point | 113 to 117 °C |
Loss on drying | ≤0.5 % |
Solubility | (2 % in water) Clear colorless solution |
HPLC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...