Carvedilol
Lieferant: Thermo Scientific
|
|
Synonyme:
1-(9H-Carbazol-4-yloxy)-3-[[2-(2-methoxyphenoxy)ethyl]amino]-2-propanol
462450010
462450050
ACRO462450010EA
33.2
EUR
ACRO462450010
ACRO462450050
Carvedilol
Carvedilol
- Bioactive
Formel:
C₂₄H₂₆N₂O₄ Molecular Weight: 406,48 g/mol Schmelzpunkt: 114…115 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00864692 CAS-Nummer: 72956-09-3 UN: 3077 ADR: 9,III |
Spezifikation Testergebnisse
Appearance | White to almost white Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 115 to 119 °C |
Loss on drying | ≤0.5 % |
HPLC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...