Lumiracoxib

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-(2-((2-chloro-6-fluorophenyl)amino)-5-methylphenyl)acetic acid, CGS-35189, COX-189,  

462801000 462802500
ACRO462801000EA 123 EUR
ACRO462801000 ACRO462802500
Lumiracoxib
Lumiracoxib

  • Bioactive

Formel: C₁₅H₁₃ClFNO₂
Molecular Weight: 293,72 g/mol
Schmelzpunkt: 139…141 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD07186254
CAS-Nummer: 220991-20-8

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow Crystalline powder
Infrared spectrum Conforms
Water ≤0.5 %
Sulfated ash ≤0.3 %
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR