(+/-)-Usnic acid

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Usnic acid, 2,6-Diacetyl-7,9-dihydroxy-8,9b-dimethyldibenzo[b,d]furan-1,3(2H,9bH)-dione

462850010 462850050 462850250
ACRO462850010EA 29.4 EUR
ACRO462850010 ACRO462850050 ACRO462850250
(+/-)-Usnic acid
(+/-)-Usnic acid

  • Bioactive

Formel: C₁₈H₁₆O₇
Molecular Weight: 344.3155 g/mol
Schmelzpunkt: 201…203 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00065294
CAS-Nummer: 125-46-2
UN: 3077
ADR: 9 III

Bestellen

Spezifikation Testergebnisse

Appearance Yellow to light green or gold Powder
Infrared spectrum Conforms
Melting point 193 to 203 °C
Loss on drying ≤2 %
Heavy metals ≤10 ppm
HPLC ≥97.5 %
Residue after ignition ≤2 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR