Tigecycline

Lieferant: Thermo Scientific
Achtung

Synonyme: 9-tert-Butylglycylamidominocycline, (4S,4aS,5aR,12aS)-9-(2-(tert-Butylamino)acetamido)-4,7-bis(dimethylamino)-3,10,12,12a-tetrahydroxy-1,11-dioxo-1,4,4a,5,5a,6,11,12a-octahydrotetracene-2-carboxamide

463060010 463062500
ACRO463060010EA 465 EUR
ACRO463060010 ACRO463062500
Tigecycline
Tigecycline

  • Bioactive

Formel: C₂₉H₃₉N₅O₈
Molecular Weight: 585,658 g/mol
Schmelzpunkt: 165…175 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00935753
CAS-Nummer: 220620-09-7

Bestellen

Spezifikation Testergebnisse

Appearance Almost white to yellow-brown Powder
Infrared spectrum Conforms
Water ≤3 %
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR