Tylosin tartrate

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Pharmasin, [R-(R'R')]-2,3-Dihydroxybutanedionate Tylosin salt

463070050 463070250 463071000
ACRO463070050EA 98.8 EUR
ACRO463070050 ACRO463070250 ACRO463071000
Tylosin tartrate
Tylosin tartrate

  • Bioactive

Formel: C₄₆H₇₇NO₁₇·C₄H₆O₆
Molecular Weight: 1066.20 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00070094
CAS-Nummer: 74610-55-2

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Powder
Infrared spectrum Conforms
Solubility (5 % in water) Clear light yellow to yellow-green
HPLC ≥94 % (sum of tylosins A+B+C+D)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR