Altretamine

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Hexamethylmelamine, NSC -13875, 2,4,6-Tris(dimethylamino)-1,3,5-triazine Hexamethylmelamine, N2,N2,N4,N4,N6,N6-hexamethyl-1,3,5-triazine-2,4,6-triamine

463090010 463090050
ACRO463090010EA 97.8 EUR
ACRO463090010 ACRO463090050
Altretamine
Altretamine

  • Bioactive

Formel: C₉H₁₈N₆
Molecular Weight: 210.28 g/mol
Schmelzpunkt: 171…175 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00549245
CAS-Nummer: 645-05-6

Bestellen

Spezifikation Testergebnisse

Appearance White to almost white Powder
Infrared spectrum Conforms
Melting point 171 to 175 °C
Loss on drying ≤1 %
HPLC ≥97.5 %

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR