Amifostine trihydrate

Lieferant: Thermo Scientific
Achtung

Synonyme: S-(2-((3-Aminopropyl)amino)ethyl) O,O-dihydrogen phosphorothioate trihydrate

463100010 463102500
ACRO463100010EA 577 EUR
ACRO463100010 ACRO463102500
Amifostine trihydrate
Amifostine trihydrate

  • Bioactive

Formel: C₅H₁₅N₂O₃PS·₃H₂O
Molecular Weight: 268,27 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00453866
CAS-Nummer: 112901-68-5

Bestellen

Spezifikation Testergebnisse

Appearance White to almost white Powder
Infrared spectrum Conforms
Water 18 to 22 %
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR