Famciclovir

Lieferant: Thermo Scientific

Synonyme: 2-[2-(2-Amino-9H-purin-9-yl)ethyl]-1,3-propanediol Diacetate, 2-(2-(2-Amino-9H-purin-9-yl)ethyl)propane-1,3-diyl diacetate

463170010 463170050
ACRO463170010EA 75.1 EUR
ACRO463170010 ACRO463170050
Famciclovir
Famciclovir

  • Bioactive

Formel: C₁₄H₁₉N₅O₄
Molecular Weight: 321,34 g/mol
Schmelzpunkt: 105 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00866964
CAS-Nummer: 104227-87-4

Bestellen

Spezifikation Testergebnisse

Appearance White to almost white Crystalline powder
Infrared spectrum Conforms
Melting point 103 to 107 °C
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR