Mosapride citrate

Lieferant: Thermo Scientific
Gefahr

Synonyme: 4-Amino-5-chloro-2-ethoxy-N-((4-(4-fluorobenzyl)morpholin-2-yl)methyl)benzamide 2-hydroxypropane-1,2,3-tricarboxylate, TAK-370, AS-4370 citrate

463200010 463200050
ACRO463200010EA 94 EUR
ACRO463200010 ACRO463200050
Mosapride citrate
Mosapride citrate

  • Bioactive

Formel: C₂₁H₂₅ClFN₃O₃·C₆H₈O₇
Molecular Weight: 614,02 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD01666680
CAS-Nummer: 112885-42-4

Bestellen

Spezifikation Testergebnisse

Appearance White to almost white powder
Infrared spectrum Conforms
Water 5 to 6 %
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR