Pramipexole dihydrochloride monohydrate ≥98%

Lieferant: Thermo Scientific

Synonyme: (S)-2-Amino-4,5,6,7-tetrahydro-6-(propylamino)benzothiazole Dihydrochloride, (S)-N6-Propyl-4,5,6,7-tetrahydrobenzo[d]thiazole-2,6-diamine dihydrochloride hydrate

463220050 463220250
ACRO463220050EA 234 EUR
ACRO463220050 ACRO463220250
Pramipexole dihydrochloride monohydrate ≥98%
Pramipexole dihydrochloride monohydrate

  • Bioactive

Formel: C₁₀H₂₁Cl₂N₃OS
Molecular Weight: 302,26 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD02183927
CAS-Nummer: 191217-81-9

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Loss on drying 4.5 % to 7 %
HPLC ≥97.5 %
Optical rotation -65° to -69° (1 % in MeOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR