Sildenafil
Lieferant: Thermo Fisher Scientific
|
Achtung
|
463240010
463242500
ACRO463240010EA
217
EUR
ACRO463240010
ACRO463242500
Sildenafil
Sildenafil
- Bioactive
Formel:
C₂₂H₃₀N₆O₄S Molecular Weight: 474.58 g/mol Schmelzpunkt: 188…190 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00867528 CAS-Nummer: 139755-83-2 |
Spezifikation Testergebnisse
Appearance | White to almost white Crystalline powder |
Infrared spectrum | Conforms |
Water | ≤2.5 % |
HPLC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...