Tolcapone

Lieferant: Thermo Scientific
Achtung

Synonyme: 3,4-Dihydroxy-4'-methyl-5-nitrobenzophenone

463280010 463282500
ACRO463280010EA 408 EUR
ACRO463280010 ACRO463282500
Tolcapone
Tolcapone

  • Bioactive

Formel: C₁₄H₁₁NO₅
Molecular Weight: 273,24 g/mol
Schmelzpunkt: 145 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00866569
CAS-Nummer: 134308-13-7
UN: 3077
ADR: 9,III

Bestellen

Spezifikation Testergebnisse

Appearance Yellow Crystalline powder3
Infrared spectrum Conforms
Water ≤0.1 %
HPLC ≥96.0 %
Residue after ignition ≤0.1 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR