Oxymetazoline hydrochloride

Lieferant: Thermo Scientific
Gefahr

Synonyme: SOxymetazoline hydrochloride, 6-tert-butyl-3-(4,5-dihydro-1H-imidazol-2-ylmethyl)-2,4-dimethylphenol hydrochloride, Afrin hydrochloride

463390050 463390250
ACRO463390050EA 104 EUR
ACRO463390050 ACRO463390250
Oxymetazoline hydrochloride
Oxymetazoline hydrochloride

  • Bioactive

Formel: C₁₆H₂₄N₂O·HCl
Molecular Weight: 296,839 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00058147
CAS-Nummer: 2315-02-8
UN: 2811
ADR: 6.1,I

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Powder
Infrared spectrum Conforms
Loss on drying ≤1 % (105°C, 3 h)
Solubility (5 % in water) Clear solution
HPLC ≥98.5 %
Residue after ignition ≤0.1 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR