Ciclopirox

Lieferant: Thermo Scientific
Achtung

Synonyme: 6-Cyclohexyl-1-hydroxy-4-methyl-2(1H)-pyridinone, 6-Cyclohexyl-1-hydroxy-4-methylpyridin-2(1H)-one

463430010 463430050
ACRO463430010EA 49.7 EUR
ACRO463430010 ACRO463430050
Ciclopirox
Ciclopirox

  • Bioactive

Formel: C₁₂H₁₇NO₂
Molecular Weight: 207,27 g/mol
Schmelzpunkt: 128…130 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00599441
CAS-Nummer: 29342-05-0

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Powder
Infrared spectrum Conforms
Melting point 140 to 145 °C
Loss on drying ≤1.5 % (vacuum)
Titration with NaOH 97.5 to 102.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR