Ciclopirox olamine
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
6-Cyclohexyl-1-hydroxy-4-methylpyridin-2(1H)-one 2-aminoethanol salt
463440010
463440050
ACRO463440010EA
45.4
EUR
ACRO463440010
ACRO463440050
Ciclopirox olamine
Ciclopirox olamine
- Bioactive
Formel:
C₁₂H₁₇NO₂·C₂H₇NO Molecular Weight: 268.3520 g/mol Schmelzpunkt: 143 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00078997 CAS-Nummer: 41621-49-2 UN: 3085 ADR: 5.1 I |
Spezifikation Testergebnisse
Appearance | White to light yellow Crystalline powder |
Infrared spectrum | Conforms |
Titration with HClO4 | 21.9 to 23.7 % (2-Aminoethanol) |
Titration with NaOH | 74.2 to 80 % (Ciclopirox) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...