(+/-)-Sulpiride
Lieferant: Thermo Scientific
Synonyme:
(+/-)-N-[(1-Ethylpyrrolidin-2-yl)methyl]-2-methoxy-5-sulphamoylbenzamide
463480050
463480250
463481000
ACRO463480050EA
42.9
EUR
ACRO463480050
ACRO463480250
ACRO463481000
(+/-)-Sulpiride
(+/-)-Sulpiride
- Bioactive
Formel:
C₁₅H₂₃N₃O₄S Molecular Weight: 341,43 g/mol Schmelzpunkt: 185 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00055061 CAS-Nummer: 15676-16-1 |
Spezifikation Testergebnisse
Appearance | White to almost white Powder and or crystals |
Infrared spectrum | Conforms |
Melting point | 177 to 181 °C |
Loss on drying | ≤0.5 % (1 g, 105 °C) |
HPLC | ≥97.5 % |
Residue after ignition | ≤0.1 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...