Oxaprozin

Lieferant: Thermo Scientific
Achtung

Synonyme: 3-(4,5-Diphenyloxazol-2-yl)propanoic acid, 4,5-Diphenyl-2-oxazolepropionic Acid

463530050 463530250
ACRO463530050EA 55.7 EUR
ACRO463530050 ACRO463530250
Oxaprozin
Oxaprozin

  • Bioactive

Formel: C₁₈H₁₅NO₃
Molecular Weight: 293,32 g/mol
Schmelzpunkt: 161…165 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00215977
CAS-Nummer: 21256-18-8
UN: 3077
ADR: 9,III

Bestellen

Spezifikation Testergebnisse

Appearance White to light beige Crystalline powder
Infrared spectrum Conforms
Melting point 161 to 165 °C
Loss on drying ≤0.3 % (105 °C, 2 h)
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR