Pseudopelletierine

Lieferant: Thermo Scientific
Achtung

Synonyme: 9-Methyl-9-azabicyclo[3.3.1]nonan-3-one

463980010 463980050 463980250
ACRO463980010EA 51.2 EUR
ACRO463980010 ACRO463980050 ACRO463980250
Pseudopelletierine
Pseudopelletierine

  • Bioactive

Formel: C₉H₁₅NO
Molecular Weight: 153,23 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00078161
CAS-Nummer: 552-70-5

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow to brown Powder
Infrared spectrum Conforms
GC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR