Vandetanib

Lieferant: Thermo Scientific

Synonyme: ZD6474, N-(4-Bromo-2-fluorophenyl)-6-methoxy-7-((1-methylpiperidin-4-yl)methoxy)quinazolin-4-amine

464330010 464330050 464332500
ACRO464330010EA 205 EUR
ACRO464330010 ACRO464330050 ACRO464332500
Vandetanib
Vandetanib

  • Bioactive

Formel: C₂₂H₂₄BrFN₄O₂
Molecular Weight: 475,4 g/mol
Schmelzpunkt: 240…243 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD07772346
CAS-Nummer: 443913-73-3

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Powder
Infrared spectrum Conforms
Loss on drying ≤0.5 %
HPLC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR