Apixaban

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: BMS56224701, 1-(4-methoxyphenyl)-7-oxo-6-[4-(2-oxopiperidin-1-yl)phenyl]-4,5-dihydropyrazolo[5,4-c]pyridine-3-carboxamide, 1-(4-Methoxyphenyl)-7-oxo-6-(4-(2-oxopiperidin-1-yl)phenyl)-4,5,6,7-tetrahydro-1H-pyrazolo[3,4-c]pyridine-3-carboxamide

464491000 464495000
ACRO464491000EA 74.4 EUR
ACRO464491000 ACRO464495000
Apixaban
Apixaban

  • Bioactive

Formel: C₂₅H₂₅N₅O₄
Molecular Weight: 459.5 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD11977295
CAS-Nummer: 503612-47-3

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 238 to 240 °C
HPLC ≥97.5 %

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR