β-L-Thymidine
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Telbivudine, 1-((2S,4R,5S)-4-Hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-methylpyrimidine-2,4(1H,3H)-dione
464550010
464550050
ACRO464550010EA
102
EUR
ACRO464550010
ACRO464550050
β-L-Thymidine
β-L-Thymidine
- Bioactive
Formel:
C₁₀H₁₄N₂O₅ Molecular Weight: 242,2286 g/mol Schmelzpunkt: 185…190 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD02683612 CAS-Nummer: 3424-98-4 |
Spezifikation Testergebnisse
Appearance | White to almost white Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 185 to 190 °C |
Loss on drying | ≤0.5 % |
HPLC | ≥98.5 % |
Residue after ignition | ≤0.2 % |
Specific optical rotation | -15° to -22° (20 °C, 589 nm) (1% in water) |
Clarity of solution | (1% in water) Clear colorless solution |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...