Ginsenoside Rb1

Lieferant: Thermo Scientific
Achtung

Synonyme: (2S,3R,4S,5S,6R)-2-(((2R,3R,4S,5S,6R)-4,5-Dihydroxy-2-(((3S,5R,8R,9R,10R,12R,13R,14R,17S)-12-hydroxy-4,4,8,10,14-pentamethyl-17-((S)-6-methyl-2-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-((((2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)methyl)tetrahydro-2H-pyran-2-yl)oxy)hept-5-en-2-yl)hexadecahydro-1H-cyclopenta[a]phenanthren-3-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol

464630500 464632500
ACRO464630500EA 105 EUR
ACRO464630500 ACRO464632500
Ginsenoside Rb1
Ginsenoside Rb1

  • Bioactive

Formel: C₅₄H₉₂O₂₃
Molecular Weight: 1109,3 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00133367
CAS-Nummer: 41753-43-9

Bestellen

Spezifikation Testergebnisse

Appearance White Powder (May have a yellow hue)
Infrared spectrum Conforms
Water ≤10 %
Solubility light yellow
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR