Glycitein

Lieferant: Thermo Scientific

Synonyme: 7-Hydroxy-3-(4-hydroxyphenyl)-6-methoxy-4H-chromen-4-one, 7,4'-Dihydroxy-6-methoxyisoflavone

464640500 464642500
ACRO464640500EA 177 EUR
ACRO464640500 ACRO464642500
Glycitein
Glycitein

  • Bioactive

Formel: C₁₆H₁₂O₅
Molecular Weight: 284,27 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00016679
CAS-Nummer: 40957-83-3

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Powder or crystals
Infrared spectrum Conforms
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR