N-Benzoyladenosin
Lieferant: Thermo Scientific
|
Achtung
|
464690010
464690050
ACRO464690010EA
75.5
EUR
ACRO464690010
ACRO464690050
N-Benzoyladenosin
N-Benzoyladenosin
- Bioactive
Formel:
C₁₇H₁₇N₅O₅ Molecular Weight: 371,35 g/mol Schmelzpunkt: 141…145 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00056988 CAS-Nummer: 4546-55-8 |
Spezifikation Testergebnisse
Appearance | White to almost white Crystalline powder or crystals |
Infrared spectrum | Conforms |
Water | ≤5 % |
HPLC | ≥95.0 % |
Specific optical rotation | -46° to -52° (20°C, 589 nm) (1% in DMSO) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...