N-Succinimidyl Acetate

Lieferant: Thermo Scientific
Achtung

Synonyme: N-Acetoxysuccinimide, Ac-Osu, Acetic Acid N-Succinimidyl Ester, 2,5-Dioxopyrrolidin-1-yl acetate

464750050 464750250 464751000
ACRO464750050EA 51.7 EUR
ACRO464750050 ACRO464750250 ACRO464751000
N-Succinimidyl Acetate
N-Succinimidyl Acetate

  • Bioactive

Formel: C₆H₇NO₄
Molecular Weight: 157,13 g/mol
Schmelzpunkt: 133…134 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00037916
CAS-Nummer: 14464-29-0

Bestellen

Spezifikation Testergebnisse

Appearance White to almost white Crystalline powder
Infrared spectrum Conforms
Melting point 132 to 136 °C
Water ≤2 %
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR