Elacridar

Lieferant: Thermo Scientific

Synonyme: N-[4-[2-(3,4-Dihydro-6,7-dimethoxy-2(1H)-isoquinolinyl)ethyl]phenyl]-9,10-dihydro-5-methoxy-9-oxo-4-acridinecarboxamide, N-(4-(2-(6,7-Dimethoxy-3,4-dihydroisoquinolin-2(1H)-yl)ethyl)phenyl)-5-methoxy-9-oxo-9,10-dihydroacridine-4-carboxamide

464950010 464952500
ACRO464950010EA 1385 EUR
ACRO464950010 ACRO464952500
Elacridar
Elacridar

  • Bioactive

Formel: C₃₄H₃₃N₃O₅
Molecular Weight: 563,654 g/mol
Schmelzpunkt: 210 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00912604
CAS-Nummer: 143664-11-3

Bestellen

Spezifikation Testergebnisse

Appearance Almost white to yellow Powder
Infrared spectrum Conforms
Water ≤1 %
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR