Schisandrin A

Lieferant: Thermo Scientific

Synonyme: (6R,7S,12aR)-5,6,7,8-Tetrahydro-1,2,3,10,11,12-hexamethoxy-6,7-dimethyldibenzo[a,c]cyclooctene, Schizandrin A, Deoxyschizandrin, Deoxyschisandrin

465030010 465032500
ACRO465030010EA 641 EUR
ACRO465030010 ACRO465032500
Schisandrin A
Schisandrin A

  • Bioactive

Formel: C₂₄H₃₂O₆
Molecular Weight: 416,51 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09026934
CAS-Nummer: 61281-38-7

Bestellen

Spezifikation Testergebnisse

Appearance White to almost white Powder
Infrared spectrum Conforms
Melting point 112 to 116 °C
Loss on drying ≤3 %
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR