Acacetin

Lieferant: Thermo Scientific
Achtung

Synonyme: 4'-Methoxy-5,7-dihydroxyflavon, Linarisenin, 5,7-Dihydroxy-2-(4-methoxyphenyl)chromen-4-on, 5,7-Dihydroxy-2-(4-methoxyphenyl)-4H-chromen-4-on, 5,7-Dihydroxy-2-(4-methoxyphenyl)-4H-1-benzopyran-4-on, 5,7-Dihydroxy-4'-methoxyflavon, Linarigenin, Buddleoflavonol, 5,7-Dihydroxy-2-(4-methoxyphenyl)-4-benzopyron, 4'-Methoxyapigenin

465230010 465232500
ACRO465230010EA 145 EUR
ACRO465230010 ACRO465232500
Acacetin
Acacetin

  • Bioactive

Formel: C₁₆H₁₂O₅
Molecular Weight: 284,27 g/mol
Schmelzpunkt: 260…265 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00016936
CAS-Nummer: 480-44-4

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow to light brown Powder or crystals
Infrared spectrum Conforms
Water ≤3 %
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR