Oxybutynin hydrochloride
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Oxybutynin chloride, 4-(Diethylamino)but-2-yn-1-yl 2-cyclohexyl-2-hydroxy-2-phenylacetate hydrochloride
463500010
463500050
463500250
ACRO463500010EA
96.9
EUR
ACRO463500010
ACRO463500050
ACRO463500250
Oxybutynin hydrochloride
Oxybutynin hydrochloride
- Bioactive
Formel:
C₂₂H₃₁NO₃·HCl Molecular Weight: 393.95 g/mol Schmelzpunkt: 125…128 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00072150 CAS-Nummer: 1508-65-2 |
Spezifikation Testergebnisse
Appearance | White to almost white Crystalline powder |
Infrared spectrum | Conforms |
Loss on drying | ≤3 % |
Titration with NaOH | 97.5 to 102.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...