Acetamiprid
Lieferant: Thermo Scientific
|
Gefahr
|
463930010
463932500
ACRO463930010EA
298
EUR
ACRO463930010
ACRO463932500
Acetamiprid
Acetamiprid
- Bioactive
Formel:
C₁₀H₁₁ClN₄ Molecular Weight: 222,67 g/mol Schmelzpunkt: 100…102 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD06201842 CAS-Nummer: 135410-20-7 UN: 2811 ADR: 6.1,II |
Spezifikation Testergebnisse
Appearance | White to light yellow Powder |
Infrared spectrum | Conforms |
Melting point | 95 to 102 °C |
Water | ≤1 % |
HPLC | ≥95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...