(-)-Scopolamine n-butylbromide
Lieferant: Thermo Fisher Scientific
|
Achtung
|
464840010
464840050
ACRO464840010EA
52.2
EUR
ACRO464840010
ACRO464840050
(-)-Scopolamine n-butylbromide
(-)-Scopolamine n-butylbromide
- Bioactive
Formel:
C₂₁H₃₀BrNO₄ Molecular Weight: 440.38 g/mol Schmelzpunkt: 142…144 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00078561 CAS-Nummer: 149-64-4 |
Spezifikation Testergebnisse
Appearance | White to almost white Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 138 to 142 °C |
Loss on drying | ≤3 % (105 °C) |
Solubility | (5% in water) Clear colorless solution |
Titration Argentimetric | 97.5 to 102.5 % |
Specific optical rotation | -16° to -22° (20 °C, 589 nm) (5% in water) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...