Retigabine dihydrochloride

Lieferant: Thermo Fisher Scientific
Achtung

465010010 465012500
ACRO465010010EA 625 EUR
ACRO465010010 ACRO465012500
Retigabine dihydrochloride
Retigabine dihydrochloride

  • Bioactive

Formel: C₁₆H₁₈FN₃O₂·₂HCl
Molecular Weight: 376.25 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04307735
CAS-Nummer: 150812-13-8

Bestellen

Spezifikation Testergebnisse

Appearance Almost white Powder
Infrared spectrum Conforms
Water ≤1 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR