Scutellarin
Lieferant: Thermo Fisher Scientific
Synonyme:
(2S,3S,4S,5R,6S)-6-((5,6-Dihydroxy-2-(4-hydroxyphenyl)-4-oxo-4H-chromen-7-yl)oxy)-3,4,5-trihydroxytetrahydro-2H-pyran-2-carboxylic acid
465130010
465132500
ACRO465130010EA
419
EUR
ACRO465130010
ACRO465132500
Scutellarin
Scutellarin
- Bioactive
Formel:
C₂₁H₁₈O₁₂ Molecular Weight: 462.36 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD01861503 CAS-Nummer: 27740-01-8 |
Spezifikation Testergebnisse
Appearance | Yellow to brown Powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...