ITIC-M, Reagenz für organische Photovoltaik (OVP)
Lieferant: Thermo Scientific
H66035.MC
H66035.MD
H66035.ME
H66035.MCEA
612
EUR
H66035.MC
H66035.MD
H66035.ME
ITIC-M, Reagenz für organische Photovoltaik (OVP)
ITIC-M
Formel:
C₉₆H₈₆O₂N₄S₄ Molecular Weight: 1456,00 g/mol |
CAS-Nummer:
2047352-80-5 |
Spezifikation Testergebnisse
Appearance (Color) | Black |
Form | Flake or threadlike |
Bandgap energy | 1,65 ± 0,02 eV |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...