IEICO-4Cl, Reagenz für organische Photovoltaik (OVP)

Lieferant: Thermo Scientific

H66752.MC H66752.MD H66752.ME
H66752.MCEA 612 EUR
H66752.MC H66752.MD H66752.ME
IEICO-4Cl, Reagenz für organische Photovoltaik (OVP)
IEICO-4Cl
Formel: C₁₁₄H₁₁₄Cl₄N₄O₄S₄
Molecular Weight: 1874,22 g/mol
CAS-Nummer: 2240998-88-1

Bestellen

Spezifikation Testergebnisse

Appearance (Color) Black
Form Flake or threadlike
Bandgap energy 1,26 ± 0,02 eV

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR