IEICO, Reagenz für organische Photovoltaik (OVP)
Lieferant: Thermo Scientific
H66656.MC
H66656.MD
H66656.ME
H66656.MCEA
630
EUR
H66656.MC
H66656.MD
H66656.ME
IEICO, Reagenz für organische Photovoltaik (OVP)
IEICO
Formel:
C₁₁₄H₁₁₈N₄O₄S₄ Molecular Weight: 1736,44 g/mol |
CAS-Nummer:
2055812-53-6 |
Spezifikation Testergebnisse
Appearance (Color) | Black |
Form | Flake or threadlike |
Bandgap energy | 1,34 ± 0,02 eV |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...