o-IDTBr, Reagenz für organische Photovoltaik (OVP)
Lieferant: Thermo Scientific
H66666.MC
H66666.MD
H66666.ME
H66666.MCEA
673
EUR
H66666.MC
H66666.MD
H66666.ME
o-IDTBr, Reagenz für organische Photovoltaik (OVP)
o-IDTBr
Formel:
C₇₂H₈₈N₆O₂S₈ Molecular Weight: 1326,03 g/mol |
CAS-Nummer:
2077945-91-4 |
Spezifikation Testergebnisse
Appearance (Color) | Black |
Form | Flake or threadlike |
Bandgap energy | 1,60 ± 0,02 eV |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...