ITIC-Cl, Reagenz für organische Photovoltaik (OVP)

Lieferant: Thermo Scientific

H66521.MC H66521.MD H66521.ME
H66521.MCEA 612 EUR
H66521.MC H66521.MD H66521.ME
ITIC-Cl, Reagenz für organische Photovoltaik (OVP)
ITIC-Cl
Formel: C₉₄H₇₈Cl₄O₂S₄
Molecular Weight: 1565,72 g/mol
CAS-Nummer: 2253663-81-7

Bestellen

Spezifikation Testergebnisse

Appearance (Color) Black
Form Flake or threadlike
Bandgap energy 1,46 ± 0,02 eV

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR