ITIC-F, Reagenz für organische Photovoltaik (OVP)

Lieferant: Thermo Scientific

H66664.MC H66664.MD H66664.ME
H66664.MCEA 612 EUR
H66664.MC H66664.MD H66664.ME
ITIC-F, Reagenz für organische Photovoltaik (OVP)
ITIC-F
Formel: C₉₄H₇₈F₄N₄O₂S₄
Molecular Weight: 1499,88 g/mol
CAS-Nummer: 2097998-59-7

Bestellen

Spezifikation Testergebnisse

Appearance (Color) Black
Form Flake or threadlike
Bandgap energy 1,55 ± 0,02 eV

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR