ITIC, Reagenz für organische Photovoltaik (OVP)

Lieferant: Thermo Scientific

H66574.MC H66574.MD H66574.ME
H66574.MCEA 489 EUR
H66574.MC H66574.MD H66574.ME
ITIC, Reagenz für organische Photovoltaik (OVP)
ITIC
Formel: C₉₄H₈₂N₄O₂S₄
Molecular Weight: 1427,94 g/mol
CAS-Nummer: 1664293-06-4

Bestellen

Spezifikation Testergebnisse

Appearance (Color) Black
Form Flake or threadlike
Bandgap energy 1,57 ± 0,02 eV

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR