Estriol 3-sulphate sodium salt ≥98% (durch TLC), Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
|
Achtung
|
Synonyme:
1,3,5(10)-Estratriene-3,16alpha,17beta-triol 3-sulphate, 3,16alpha,17beta-Trihydroxy-1,3,5(10)-estratriene 3-sulphate
M-Clarity™ Qualitätsstufe = MQ200
E6375-10MG
E6375-25MG
E6375-10MGEA
534
EUR
E6375-10MG
E6375-25MG
Estriol 3-sulphate sodium salt ≥98% (durch TLC), Sigma-Aldrich®
Estriol 3-sulphate sodium salt
Estriol 3-sulfate sodium salt, (1,3,5(10)-Estratriene-3,16alpha,17beta-triol 3-sulfate; 3,16alpha,17beta-Trihydroxy-1,3,5(10)-estratriene 3-sulfate)
Verpackung: Package size based on steroid content.
Formel:
C₁₈H₂₃NaO₆S Molecular Weight: 390,43 g/mol Lagertemperatur: Room Temperature |
MDL:
MFCD00056547 CAS-Nummer: 5150-64-1 |
Spezifikation Testergebnisse
Appearance (Form) | Powder or Lyophilised Powder |
Appearance (Colour) | White to Off-White |
Solubility (Solvent) | Methanol |
Solubility (Conc) | 19.60 - 20.40 mg/ml |
Solubility (Turbidity) | Clear |
Solubility (Colour) | Colorless to Faint Yellow |
Purity (TLC) | ≥98.00 % |
Stabiliser Content(Stabiliser) | Methyl Glucamine |
Stabiliser Content (Method) | UV |
Stabiliser Content (%) | 25.00 - 35.00 % |
Storage Conditions | Under Argon |
Recommended Retest Period | 5 Years |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...