2-[4-(Dimethylamino)styryl]-N-methylbenzoxazolium perchlorate, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Achtung

D1429-25MG D1429-5MG
D1429-25MGEA 669 EUR
D1429-25MG D1429-5MG
2-[4-(Dimethylamino)styryl]-N-methylbenzoxazolium perchlorate, Sigma-Aldrich®
2-[4-(Dimethylamino)styryl]-N-methylbenzoxazolium perchlorate

DNA, RNA stain with advantageous spectral properties for cytometric applications.

Formel: C₁₈H₁₉ClN₂O₅
Molecular Weight: 378.81 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00274415
CAS-Nummer: 64872-13-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9e076627df0aff8fc11032ae6603d1c42d3a3be2&vid=b2bd54af8591160a7067e7ba087e2df054fc25e9&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR