4-Formylbenzene-1,3-disulphonic acid disodium salt hydrate 97%, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Achtung

Synonyme: Sodium 4-formylbenzene-1,3-disulphonate, Benzaldehyde 2,4-disulphonic acid disodium salt

M-Clarity™ Qualitätsstufe = MQ200

230901-10G 230901-50G
230901-10GEA 112 EUR
230901-10G 230901-50G
4-Formylbenzene-1,3-disulphonic acid disodium salt hydrate 97%, Sigma-Aldrich®
4-Formylbenzene-1,3-disulphonic acid disodium salt hydrate

4-Formylbenzene-1,3-disulfonic acid disodium salt hydrate,97% (Benzaldehyde 2,4-disulfonic acid disodium salt; Sodium 4-formylbenzene-1,3-disulfonate).


4-Formylbenzene-1,3-disulfonic acid disodium salt hydrate is used in the synthesis of polymer electrolyte membranes, which forms a crucial component of electrochemical devices.

Formel: C₇H₄Na₂O₇S₂
Lagertemperatur: Room Temperature
MDL: MFCD00150711
CAS-Nummer: 207291-88-1
EINECS: 251-551-0

Bestellen

Spezifikation Testergebnisse

Appearance (Colour) White to Yellow
Appearance (Form): Powder, Crystals, Crystalline Powder and/or Chunks Conforms to Requirements
Infrared spectrum Conforms to Structure
Water (by Karl Fischer) Typically 2 - 16%
Purity (HPLC) ≥96.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR