Kinyoun's carbol fuchsin, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
|
Gefahr
|
Synonyme:
Dimethyl fuchsin, Carbol-fuchsin, alpha4-(p-Aminophenyl)-alpha4-(4-imino-3-methyl-2,5-cyclohexadien-1-ylidene)-2,4-xylidine monohydrochloride
21819-250ML-F
21819-50ML-F
21819-250ML-FEA
432
EUR
21819-250ML-F
21819-50ML-F
Kinyoun's carbol fuchsin, Sigma-Aldrich®
Kinyoun's carbol fuchsin
Carbol-Fuchsin solution according to Kinyoun,for microscopy (Kinyoun's carbol-Fuchsin)
Formel:
C₂₁H₂₂ClN₃ Molecular Weight: 351,878 g/mol Dichte: 0,99 g/mL (20 °C) Lagertemperatur: Raumtemperatur |
CAS-Nummer:
26261-57-4 ADR: 3,3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...