4-(Dimethylamino)azobenzene-4'-isothiocyanate 97%, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
|
Gefahr
|
Synonyme:
DABITC, 4-(N,N-Dimethylamino)azobenzene-4'-isothiocyanate, 4-(4-Isothiocyanatophenylazo)-N,N-dimethylaniline, 4-((4-Isothiocyanatophenyl)diazenyl)-N,N-dimethylaniline
317802-1G
317802-250MG
317802-1GEA
555
EUR
317802-1G
317802-250MG
4-(Dimethylamino)azobenzene-4'-isothiocyanate 97%, Sigma-Aldrich®
4-(Dimethylamino)azobenzene-4'-isothiocyanate
4-(4-Isothiocyanatophenylazo)-N,N-dimethylaniline,97% (4-(Dimethylamino)azobenzene-4'-isothiocyanate; DABITC)
Formel:
C₁₅H₁₄N₄S Molecular Weight: 282,37 g/mol Schmelzpunkt: 167…171 °C (Lit.) Lagertemperatur: Room Temperature |
MDL:
MFCD00004812 CAS-Nummer: 7612-98-8 |
Spezifikation Testergebnisse
Appearance (Colour) |
Conforms to Requirements Orange to Red-Brown |
Appearance (Form) |
Conforms to Requirements Powder or Crystals |
Infrared spectrum |
Conforms to Structure Orange to Red-Brown |
Carbon | 61.6 - 66.0 % |
Nitrogen | 19.1 - 20.5 % |
Purity (HPLC) | ≥96.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...